missing translation for 'onlineSavingsMsg'
Learn More

Iminostilbene, 97%

Catalog No. AC162541000
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162541000 100 g Glass bottle
AC162540100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162541000 Supplier Thermo Scientific Chemicals Supplier No. 162541000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 256-96-2 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 256-96-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00005071
InChI Key LCGTWRLJTMHIQZ-UHFFFAOYSA-N
Synonym iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-imino
PubChem CID 9212
ChEBI CHEBI:47802
IUPAC Name 11H-benzo[b][1]benzazepine
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2

Specifications

Melting Point 197°C to 201°C
Color Orange-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 193.25
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material Iminostilbene, 97%
EINECSNumber 205-970-0

RUO – Research Use Only