missing translation for 'onlineSavingsMsg'
Learn More

Iminostilbene, 97%

Catalog No. AAB2132336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2132336 500 g
AAB2132314 25 g
AAB2132322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2132336 Supplier Thermo Scientific Chemicals Supplier No. B2132336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 256-96-2 | C14H11N | 193.249 g/mol

Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminostilbene has good antioxidant effects. N-acetyl iminostilbene was synthesized by incubating iminostilbene (2.0020 g, 10.360 mmol) with acetic anhydride.

Solubility
Soluble in ethyl acetate (25 mg/ml), water (partly), dioxane, chloroform, and DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 256-96-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00005071
InChI Key LCGTWRLJTMHIQZ-UHFFFAOYSA-N
Synonym iminostilbene, 5h-dibenzo b,f azepine, 5h-dibenz b,f azepine, 2,2'-iminostilbene, dibenz b,f azepine, 2,3,6,7-dibenzazepine, stilbene, 2,2'-imino
PubChem CID 9212
ChEBI CHEBI:47802
IUPAC Name 11H-benzo[b][1]benzazepine
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C3N2

Specifications

Melting Point 197°C to 199°C
Quantity 500 g
Beilstein 1343358
Solubility Information Soluble in ethyl acetate (25mg/ml),water (partly),dioxane,chloroform,and DMSO.
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material Iminostilbene
EINECSNumber 205-970-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only