missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidurea, 26-28% nitrogen
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| pH | 6.0 to 7.5 (1% aq. soln.) |
| Loss on Drying | 3.0 % max. (48 hrs) (vacuum, P2O5) |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Merck Index | 15, 4957 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 388.29 |
| Percent Purity | 26-28% nitrogen |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. |
| EINECSNumber | 254-372-6 |
| RTECSNumber | YT7365000 |
| TSCA | TSCA |
RUO – Research Use Only