Learn More
Imidazolidinyl urea
CAS: 39236-46-9 | C11H16N8O8 | 388.297 g/mol
Supplier: Thermo Scientific Chemicals H6072922
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Imidazolidinyl urea | |
| 141°C to 143°C | |
| MFCD00038890 | |
| imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea | |
| C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO | |
| 388.297 | |
| CHEBI:51805 |
| 39236-46-9 | |
| C11H16N8O8 | |
| 100 g | |
| ZCTXEAQXZGPWFG-UHFFFAOYSA-N | |
| 1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea | |
| 38258 | |
| 388.3 |
Chemical Identifiers
| 39236-46-9 | |
| 388.297 | |
| ZCTXEAQXZGPWFG-UHFFFAOYSA-N | |
| 38258 | |
| 1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea |
| C11H16N8O8 | |
| MFCD00038890 | |
| imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea | |
| CHEBI:51805 | |
| C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
EINECSNumber : 254-372-6
RTECSNumber : YT7365000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only