missing translation for 'onlineSavingsMsg'
Learn More

Imidazolidinyl urea

Catalog No. AAH6072914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6072914 25 g
AAH6072922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6072914 Supplier Thermo Scientific Chemicals Supplier No. H6072914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39236-46-9 | C11H16N8O8 | 388.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39236-46-9
Molecular Formula C11H16N8O8
Molecular Weight (g/mol) 388.297
MDL Number MFCD00038890
InChI Key ZCTXEAQXZGPWFG-UHFFFAOYSA-N
Synonym imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea
PubChem CID 38258
ChEBI CHEBI:51805
IUPAC Name 1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
SMILES C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO

Specifications

Melting Point 141°C to 143°C
Quantity 25 g
Formula Weight 388.3
Chemical Name or Material Imidazolidinyl urea
Hazard Category H317
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 254-372-6
RTECSNumber YT7365000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only