missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazolidinyl Urea 90.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 388.30 |
| Percent Purity | ≥90.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Imidazolidinyl Urea |
Safety and Handling
| RTECSNumber | YT7365000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |