missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Imidazole, ULTROL™ Grade, Calbiochem™,

Catalog No. 4015100gm
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4015100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4015100GM Supplier MilliporeSigma™ Supplier No. 4015100GM
Only null left

Useful in the pH range of 6.2 to 7.8

Specifications

CAS 288-32-4
Quantity 100 g
Molecular Formula C3H4N2
MDL Number MFCD00005183
Synonym 1,3-Diaza-2,4-cyclopentadiene
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.079
Grade ULTROL™ Grade