missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019564
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019564 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019564 Supplier Merck Emd Millipore Supplier No. PHR1180500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 288-32-4
Vapor Pressure <1 mmHg (20°C)
Physical Form Neat
Linear Formula C3H4N2
MDL Number MFCD00005183
Melting Point 88°C to 91°C
Boiling Point 256°C
Synonym 1,3-Diaza-2,4-cyclopentadiene; Glyoxaline
Quantity 500 mg
Formula Weight 68.08
Grade Certified Reference Material
Show More Show Less