missing translation for 'onlineSavingsMsg'
Learn More

Imidazole Hydroiodide (Low water content) 98.0+%, TCI America™

Catalog No. p-7131688
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09701G 1 g
I09705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09701G Supplier TCI America Supplier No. I09701G
Only null left

Chemical Identifiers

CAS 68007-08-9
Molecular Formula C3H5IN2
Molecular Weight (g/mol) 195.991
InChI Key JBOIAZWJIACNJF-UHFFFAOYSA-N
Synonym Imidazolium Iodide
PubChem CID 12203051
IUPAC Name 1H-imidazole;hydroiodide
SMILES C1=CN=CN1.I

Specifications

Quantity 1 g
Formula Weight 195.99
Percent Purity ≥98.0% (T)
Chemical Name or Material Imidazole Hydroiodide (Low water content)