missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imidazole, ACS, 99+%

Catalog No. AA4727422
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4727422 100 g
AA4727436 500 g
AA472740E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4727422 Supplier Thermo Scientific™ Supplier No. 4727422

Specifications

CAS 288-32-4
Melting Point 88-92°
Density 1.03
Boiling Point 256°
Flash Point 145°(293°F)
Molecular Formula C{3}H{4}N{2}
Quantity 100 g
MDL Number MFCD00005183
UN Number UN3263
Merck Index 14,4912
Synonym Glyoxaline;
Formula Weight 68.08
Percent Purity 99+%
Physical Form Solid
Show More Show Less