missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-4-carboxaldehyde, 97%

Catalog No. AAB2505003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2505003 1 g
AAB2505006 5 g
AAB2505014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2505003 Supplier Thermo Scientific Chemicals Supplier No. B2505003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3034-50-2 | C4H4N2O | 96.089 g/mol

Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole-4-carboxaldehyde is a 4-formyl derivative of imidazole used in the preparation of C17,20-lyase inhibitor for the treatment of androgen-dependent prostate cancer. It is also used in the synthesis of other biologically active compounds such as antimalarial drugs, fabrication of colorimetric chemosensor.

Solubility
Soluble in DMSO, methanol.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 3034-50-2
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.089
MDL Number MFCD00173726
InChI Key ZQEXIXXJFSQPNA-UHFFFAOYSA-N
Synonym 1h-imidazole-4-carbaldehyde, imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, imidazole-4-carboxaldehyde, 4-imidazolecarboxaldehyde, 4-formylimidazole, 4 5-imidazolecarboxaldehyde, 3h-imidazole-4-carbaldehyde, 4-formyl-1h-imidazole, imidazole-5-carbaldehyde
PubChem CID 76428
IUPAC Name 1H-imidazole-5-carbaldehyde
SMILES C1=C(NC=N1)C=O

Specifications

Melting Point 174°C to 177°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in DMSO,methanol.
Formula Weight 96.09
Percent Purity 97%
Chemical Name or Material Imidazole-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-227-3
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only