missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-4(5)-acetic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. p-7131285
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02291G 1 g
I02295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I02291G Supplier TCI America Supplier No. I02291G

Chemical Identifiers

CAS 3251-69-2
Molecular Formula C5H7ClN2O2
Molecular Weight (g/mol) 162.57
MDL Number MFCD00012698
InChI Key MWHLCFYPFGFBQO-UHFFFAOYSA-N
Synonym 4-imidazoleacetic acid hydrochloride, 2-1h-imidazol-5-yl acetic acid hydrochloride, imidazole-4-acetic acid hydrochloride, 2-1h-imidazol-4-yl acetic acid hydrochloride, 4-imidazoleacetic acid hcl, imidazoleacetic acid hydrochloride, unii-et7a74roao, 4-imidazoleaceticacidhydrochloride, i4aa, et7a74roao
PubChem CID 145685
IUPAC Name 2-(1H-imidazol-5-yl)acetic acid hydrochloride
SMILES Cl.OC(=O)CC1=CN=CN1

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 1 g
Formula Weight 162.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Imidazole-4(5)-acetic Acid Hydrochloride