missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. I08095G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08095G 5 g
I08091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08095G Supplier TCI America Supplier No. I08095G

Chemical Identifiers

CAS 10111-08-7
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003544
InChI Key XYHKNCXZYYTLRG-UHFFFAOYSA-N
Synonym imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole
PubChem CID 24955
IUPAC Name 1H-imidazole-2-carbaldehyde
SMILES O=CC1=NC=CN1

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 5 g
Formula Weight 96.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Imidazole-2-carboxaldehyde
TSCA No
Recommended Storage Refrigerator