missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-2-carboxaldehyde, 97%

Catalog No. AAL1388306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1388306 5 g
AAL1388314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1388306 Supplier Thermo Scientific Chemicals Supplier No. L1388306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10111-08-7 | C4H4N2O | 96.09 g/mol

Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.

Solubility
Soluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10111-08-7
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003544
InChI Key XYHKNCXZYYTLRG-UHFFFAOYSA-N
Synonym imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole
PubChem CID 24955
IUPAC Name 1H-imidazole-2-carbaldehyde
SMILES O=CC1=NC=CN1

Specifications

Melting Point ∽205°C (decomposition)
Quantity 5 g
Beilstein 4371302
Sensitivity Air sensitive
Solubility Information Soluble in water.
Formula Weight 96.09
Percent Purity 97%
Chemical Name or Material Imidazole-2-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only