missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-b]pyridazine 98.0+%, TCI America™

Catalog No. I07421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07421G 1 g
I07425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07421G Supplier TCI America Supplier No. I07421G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 766-55-2
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD07782103
InChI Key VTVRXITWWZGKHV-UHFFFAOYSA-N
Synonym imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
PubChem CID 136599
IUPAC Name imidazo[1,2-b]pyridazine
SMILES C1=CC2=NC=CN2N=C1

Specifications

Melting Point 54°C
Quantity 1 g
Formula Weight 119.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Imidazo[1,2-b]pyridazine