missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride, 97%

Catalog No. AC453360010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453360010 1 g
AC453360050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453360010 Supplier Thermo Scientific Chemicals Supplier No. 453360010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1423031-35-9 | C8H6N2O2·ClH | 198.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1423031-35-9
Molecular Formula C8H6N2O2·ClH
Molecular Weight (g/mol) 198.61
InChI Key OQGRMPZJJDBYBL-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-7-carboxylic acid hydrochloride, imidazo 1,2-a pyridine-7-carboxylic acid hydrochloride 1:x
PubChem CID 71756722
IUPAC Name imidazo[1,2-a]pyridine-7-carboxylic acid;hydrochloride
SMILES C1=CN2C=CN=C2C=C1C(=O)O.Cl

Specifications

Melting Point 295.0°C to 298.0°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 198.61
Percent Purity 97%
Chemical Name or Material Imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only