missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carbonitrile, ≥97%, Thermo Scientific™

Catalog No. p-3110989
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC33116CB 250mg
CC33116DA 1g
CC33116DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC33116CB Supplier Thermo Scientific Chemicals Supplier No. CC33116CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106850-34-4
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.149
MDL Number MFCD06659603
InChI Key LRJOKNYELSECDZ-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine, 4-hydroimidazo 1,2-a pyridine-6-carbonitrile, 6-cyano-imidazo 1,2-a pyridine, acmc-1bu34, 6-imidazo 1,2-a pyridinecarbonitrile, imidazo 1,2-, a pyridine-6-carbonitrile, imidazo 1,2-alpha pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine-6-carbonitrile
PubChem CID 2764471
IUPAC Name imidazo[1,2-a]pyridine-6-carbonitrile
SMILES C1=CC2=NC=CN2C=C1C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 143.15
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carbonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.