missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidapril Hydrochloride 98.0+%, TCI America™
Supplier: TCI America I0901100MG
Specifications
| Imidapril Hydrochloride | |
| 206°C | |
| 2 (1% H2O soln.) | |
| MFCD00923828 | |
| (4S)-3-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1-methyl-2-oxo-4-imidazolidinecarboxylic Acid Hydrochloride | |
| CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2C(CN(C2=O)C)C(=O)O.Cl | |
| 441.909 | |
| 441.91 | |
| Crystalline Powder |
| 89396-94-1 | |
| White | |
| C20H28ClN3O6 | |
| 100 mg | |
| LSLQGMMMRMDXHN-GEUPQXMHSA-N | |
| (4S)-3-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-1-methyl-2-oxoimidazolidine-4-carboxylic acid;hydrochloride | |
| 5485193 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 89396-94-1 | |
| 441.909 | |
| LSLQGMMMRMDXHN-GEUPQXMHSA-N | |
| 5485193 | |
| CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2C(CN(C2=O)C)C(=O)O.Cl |
| C20H28ClN3O6 | |
| MFCD00923828 | |
| (4S)-3-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1-methyl-2-oxo-4-imidazolidinecarboxylic Acid Hydrochloride | |
| (4S)-3-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-1-methyl-2-oxoimidazolidine-4-carboxylic acid;hydrochloride |
Safety and Handling
RTECSNumber : NI8927400
TSCA : No