missing translation for 'onlineSavingsMsg'
Learn More

Imidapril Hydrochloride 98.0+%, TCI America™

Catalog No. I090125MG
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I090125MG 25 mg
I0901100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. I090125MG missing translation for 'mfr' TCI America missing translation for 'supplierNo' I090125MG
Only null left

Chemical Identifiers

CAS 89396-94-1
Molecular Formula C20H28ClN3O6
Molecular Weight (g/mol) 441.909
MDL Number MFCD00923828
InChI Key LSLQGMMMRMDXHN-GEUPQXMHSA-N
Synonym (4S)-3-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1-methyl-2-oxo-4-imidazolidinecarboxylic Acid Hydrochloride
PubChem CID 5485193
IUPAC Name (4S)-3-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-1-methyl-2-oxoimidazolidine-4-carboxylic acid;hydrochloride
SMILES CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2C(CN(C2=O)C)C(=O)O.Cl

Specifications

Melting Point 206°C
Color White
pH 2 (1% H2O soln.)
Quantity 25 mg
Formula Weight 441.91
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Imidapril Hydrochloride
RTECSNumber NI8927400
TSCA No