missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imatinib mesylate, 98%

Catalog No. AC461080010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461080010 1 g
AC461080050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461080010 Supplier Thermo Scientific Chemicals Supplier No. 461080010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 220127-57-1 | C30H35N7O4S | 589.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 220127-57-1
Molecular Formula C30H35N7O4S
Molecular Weight (g/mol) 589.72
MDL Number MFCD04307699
InChI Key YLMAHDNUQAMNNX-UHFFFAOYSA-N
Synonym imatinib mesylate, gleevec, glivec, imatinib mesilate, imatinib methanesulfonate, imatinib mesylate sti571, imatinib monomesylate, unii-8a1o1m485b, imatinib, methanesulfonate salt
PubChem CID 123596
ChEBI CHEBI:31690
IUPAC Name methanesulfonic acid;4-[(4-methylpiperazin-1-yl)methyl]-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
SMILES CS(O)(=O)=O.CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1

Specifications

Melting Point 209.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 589.71
Percent Purity 98%
Chemical Name or Material Imatinib mesylate

RUO – Research Use Only