missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Imaging System Controller, Standard

Catalog No. BT1900008
Change view
Click to view available options
Imaging System Controller; PC: ProDesk 400 G7 Small Form Factor Business PC; Includes: Keyboard, Mouse, USB 3.1 Ports, USB 2.0 Ports, Serial Port, HMDI 2.0 Port, RJ-45 Network Port, Dual-mode Display Port, VGA Port
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BT1900008 Imaging System Controller; PC: ProDesk 400 G7 Small Form Factor Business PC; Includes: Keyboard, Mouse, USB 3.1 Ports, USB 2.0 Ports, Serial Port, HMDI 2.0 Port, RJ-45 Network Port, Dual-mode Display Port, VGA Port
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BT1900008 Supplier Agilent BioTek Supplier No. 1900008S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standard Imaging System Controller is designed for intensive imaging and analysis operations.

  • Operating System: Microsoft Windows 10 Pro 64-Bit, MS Windows 11 Pro 64-Bit (v21H2)
  • Processor: Intel 9500 Core i5-6Core (6 Core, 6 Threads, 3 GHz, 9MB Cache)
  • Memory: 8 GB DDR4 2666 (1x8 GB)
  • Wireless: Intergrated Intel 9560 ac 2x2 nvP +Bluetooth 5
  • Storage: 512GB PCIe NVMe TLC SSD
  • Input Device: USB Business Slim Keyboard and Optical Mouse
  • Power: 180W Internal Power Supply
  • Ports and Connectors: (4) USB 3.1, (2) USB 2.0, (2) USB 2.0 - (Keyboard and Mouse), Serial Port, HMDI 2.0, RJ-45 Network, Dual-Mode DisplayPort, VGA
  • Ideal for applications such as: Large montages, including single-sample applications; Complex image analysis on microplate-based assays; High-object count applications like spot counting; Higher-throughput imaging application using an Agilent BioTek BioStack microplate stacker or an Agilent BioTek BioSpa automated incubator

Specifications

Description Agilent Standard Controller
For Use With (Application) Intensive Imaging and Analysis
Color Black
Includes Keyboard, Mouse, USB 3.1 Port (4), USB 2.0 Port (2), Keyboard/Mouse USB 2.0 Port (2), Serial Port (1), HMDI 2.0 Port (1), RJ-45 Network Port (1), Dual-mode Display Port (1), VGA Port (1)
Data Entry USB Business Slim Keyboard and Optical Mouse
Display 24 in. LED Monitor
Memory 8 GB DDR4 2666 (1x8 GB)
Operating System MS Windows 10 Pro 64-Bit, MS Windows 11 Pro 64-Bit (v21H2)
PC ProDesk 400 G7 Small Form Factor Business PC
Wattage 180W Internal Power Supply