missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-iT™ Plant Tissue Clearing Reagent

Catalog No. V11328
Encompass
Change view
Click to view available options
Capacity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
V11328 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V11328 Supplier Invitrogen™ Supplier No. V11328

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Image-iT Plant Tissue Clearing Reagent is used to render whole-plant tissue optically clear for imaging. Image-iT Plant Tissue Hard-Set Mountant can then be used to mount and preserve the optically cleared whole-plant tissue for imaging, storage, and archiving.

These reagents are compatible with both fluorescence-based and colorimetric-based applications. Image-iT Plant Tissue Clearing Reagent is an effective replacement for chloral hydrate, traditionally used to clear plant tissues in botanical microscopy.

Features of Image-iT Plant Tissue Clearing Reagent include:
• Clears plant tissue for whole-specimen imaging
• Soft-set mountant for imaging with refractive index of 1.45
• Effective replacement for chloral hydrate

Plant tissues often have structures with different refractive indexes, causing light scattering and spherical aberration, resulting in opaqueness. Image-iT Plant Tissue Clearing Reagent, with a high refractive index of 1.45, allows light to pass through the plant tissues with minimal light scattering, resulting in optical clarity and the ability to image deeper within the tissue.

Traditionally, researchers used chloral hydrate to clear plant specimens, but under current regulations it is a Federally Regulated Schedule IV substance (1), making it harder to acquire and use. Image-iT Plant Tissue Clearing Reagent is an effective replacement for chloral hydrate in almost all applications and tissue types where optical clearing is required.

Image-iT Plant Tissue Clearing Reagent and Image-iT Plant Tissue Hard-Set Mountant are based on Visikol™ for Plant Biology™ and Visikol™ Mount. Visikol for Plant Biology has been used in various fluorescent and colorimetric studies with plants (2,3,4) and has proved an effective substitute for chloral hydrate (1,5).

References:
1. Villani TS, Koroch AR, Simon JE. An improved clearing and mounting solution to replace chloral hydrate in microscopic applications. Appl Plant Sci. 2013 1(5).
2. Aoki Y, Van Trung N, Suzuki S. Impact of Piper betel leaf extract on grape downy mildew: effects of combining 4-allylpyrocatechol with eugenol, α-pinene or β-pinene. Plant Protect Sci. 2018 55: 23-30.
3. Lee KJI, Calder GM, Hindle CR, Newman JL, Robinson SN, Avondo JJHY, Coen ES. Macro optical projection tomography for large scale 3D imaging of plant structures and gene activity. J Exp Bot. 2017 68(3):527-538.
4. Vergara-Díaz O, Chairi F, Vicente R, Fernandez-Gallego JA, Nieto-Taladriz MT, Aparicio N, Kefauver SC, Araus JL. Leaf dorsoventrality as a paramount factor determining spectral performance in field-grown wheat under contrasting water regimes. J Exp Bot. 2018 69(12):3081-3094.
5. Urban MA, Barclay RS, Sivaguru M, Punyasena SW. Cuticle and subsurface ornamentation of intact plant leaf epidermis under confocal and superresolution microscopy. Microsc Res Tech. 2018 81(2):129-140.

Specifications

Capacity 30 mL
Content And Storage Store at 15°C to 25°C. Do not freeze.
When stored as directed, product is stable for (at least 6 months after receipt.

)
Product Line Image-iT
Product Type Tissue Clearing Reagent
Quantity 30 mL
Species Plant
Shipping Condition Room Temperature
Can I remove Image-iT Plant Tissue Hard-Set Mountant from my sample once cured?

Once cured, Image-iT Plant Tissue Hard-Set Mountant cannot be removed from the sample.

I accidentally froze Image-iT Plant Tissue Hard-Set Mountant. Can I still use it?

We do not recommend using Image-iT Plant Tissue Hard-Set Mountant that has been frozen.

How should I store samples mounted in Image-iT Plant Tissue Hard-Set Mountant?

We recommend storing the slides at room temperature in a moisture-free environment.

How long can I store samples mounted in Image-iT Plant Tissue Hard-Set Mountant?

Slides can be mounted permanently by drying excess Image-iT Plant Tissue Clearing Reagent with a lab tissue and applying Image-iT Plant Tissue Hard-Set Mountant.

I accidentally froze Image-iT Plant Tissue Clearing Reagent. Can I still use it?

We do not recommend using Image-iT Plant Tissue Clearing Reagent that has been frozen.

Do I have to use Image-iT Plant Tissue Clearing Reagent under a chemical safety hood?

We recommend following the usage guidelines specified in the MSDS.


For Research Use Only. Not for use in diagnostic procedures.