missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Solution dNTPs: DNA Polymerization Mix
Free from DNase, RNase, and nicking enzyme activity
$229.00 - $593.00
Specifications
| Format | Liquid |
|---|---|
| Concentration | 20 mM |
| Purity | >99% |
| Color | Colorless |
| For Use With (Application) | Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
45-001-458
|
Cytiva
28406558 |
4 x 10 μm |
Each for $593.00
|
|
|||||
|
45-001-457
|
Cytiva
28406557 |
10 μm |
Each for $229.00
|
|
|||||
Description
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
| Liquid | |
| >99% | |
| Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |
| 20 mM | |
| Colorless |