Learn More
Laxco™ iLED Series LED Light Source
GREENER_CHOICE
Laxco™ iLED Series boost bright LED illumination in small, compact, small foot print system.
Supplier:  Laxco™ AMPSILED23
 View more versions of this product
Catalog No. AMPSILED23
Only null left
Description
Description
Laxco™ iLED Series boost bright LED illumination in small, compact, small foot print system.
  • 12 LED strip creates a homogenized transmitted illumination
  • Dual directional upper light guides produce 720cd of focused LED illumination comparable to a 150w fiber optic light source
  • Independent variable intensity controls for both upper and lower illumination
  • 50,000 hour bulb life
  • Weighted base for stand-alone option
Certifications
Quality Proformance Certification
Warranty
2 Year - Electronic, Mechanical, Optical with option to extend to 5 years
Specifications
Chemical Identifiers
5 years optical and mechanical, 1 year electrical from date of purchase
Specifications
Weighted Base
Manufacturing, Research, Laboratory
150w equivalent LED
5 years optical and mechanical, 1 year electrical from date of purchase
32.2 x 17.1 x 37.5 cm
Power supply, Weighted base, Rechargeable battery pack, Light guide
110 to 220 V Auto Switch, External, 5 V/2 A
Product Suggestions
Product Suggestions