missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILC Dover Sentinel XL Breathing Tube
Description
- Latex free
- For use with Sentinel XL Blower System
Specifications
Specifications
| Certifications/Compliance | NIOSH TC-21C-0801 NIOSH TC-21C-0814 |
| Color | White |
| Product Type | Breathing Tube |
| For Use With (Equipment) | Sentinel XL HP System |
| Includes | 5 breathing tubes |
| Model | S-4005-5 |