missing translation for 'onlineSavingsMsg'
Learn More

ILC Dover HEPA (HP) P100 Cartridge Filters, 25 sets of 2

Catalog No. 19631051
Change view
Click to view available options
:
Includes 50 cartridges and plug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19631051 Includes 50 cartridges and plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19631051 Supplier ILC Dover Supplier No. S 4002 25
Only null left

The high efficiency particulate air cartridge filter can remove at least 99.97% of dust, pollen, mold, bacteria, and any airborne particles with a size of 0.3 microns.

  • Can remove at least 99.97% of dust, pollen, mold, bacteria, and any airborne particles with a size of 0.3 microns

Specifications

Cartridge Type HEPA Cartridge
Certifications/Compliance NIOSH
Color Magenta
Product Type Combination Cartridge/Filter
For Use With (Equipment) Powered air purifying respirator
Includes 50 cartridges and plug
Quantity 50/Pk.