Learn More
ACROBiosystems Inc ACROBiosystems Human IL-37 / IL-1F7b Protein, His Tag
Catalog No. 501957237
Encompass
Recombinant Protein;500UG;Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contains AA Val 46 - Asp 218 (Accession # AAH20637).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.3 kDa. The protein migrates as 20.3 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 501957237 Supplier ACROBiosystems Inc Supplier No. ILBH5141500UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;500UG;Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contains AA Val 46 - Asp 218 (Accession # AAH20637).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.3 kDa. The protein migrates as 20.3 kDa under reducing (R) condition (SDS-PAGE).