missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ IL-18 R beta/IL-1 R7/ACPL Antibody Blocking Peptide

Catalog No. NB398712
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398712 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398712 Supplier R&D Systems™ Supplier No. NB10056433PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems.

Specifications

Gene ID (Entrez) 8807
Product Line Novus
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-56433
Gene Alias Accessory protein-like, ACPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-18 receptor accessory protein, IL-18 receptor beta, IL-18 RAcP, IL18RB, IL-18 Rbeta, IL-18 R-beta, IL-1 R accessory protein-like, IL1R7, IL-1R7, IL-1 R-7, IL-1 RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590
Gene Symbol IL18RAP
Quantity 0.1 mg
Research Category CD Markers
Storage Buffer Peptide dissolved in dH20. Contains no BSA.