Learn More
ACROBiosystems Inc ACROBiosystems Biotinylated Human IL-15 Protein, Fc,Avitag™
Catalog No. 501957037
Encompass
Recombinant Protein;200UG;Biotinylated Human IL-15, Fc,Avitag (IL5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P40933-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) at 2 g/mL (100 L/well) can bind Biotinylated Human IL-15, Fc,Avitag (Cat. No. IL5-H82F3) with a linear range of 2-78 ng/mL (QC tested).
Catalog No. 501957037 Supplier ACROBiosystems Inc Supplier No. IL5H82F3200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;200UG;Biotinylated Human IL-15, Fc,Avitag (IL5-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P40933-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-15 R alpha, Fc Tag (Cat. No. ILA-H5253) at 2 g/mL (100 L/well) can bind Biotinylated Human IL-15, Fc,Avitag (Cat. No. IL5-H82F3) with a linear range of 2-78 ng/mL (QC tested).