missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, Granular, FCC, 99-101%, Spectrum™ Chemical

Catalog No. 18606272
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606272 50 kg Poly Bottle
18606269 500 g Poly Bottle
18606270 2.5 kg Poly Bottle
18606271 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606272 Supplier Spectrum Chemical Mfg Cor Supplier No. S111450KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1114, 6131-90-4, NaC2H3O2·3H2O

Spectrum™ Chemical Sodium Acetate, Trihydrate, Granular, FCC is used as food flavorant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.08
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
IUPAC Name sodium acetate trihydrate
SMILES O.O.O.[Na+].CC([O-])=O

Specifications

CAS Max % 1
Loss on Drying 36-41%
Packaging Poly Bottle
Quantity 50 kg
Percent Purity 99.0 to 101%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sodium Acetate, Trihydrate, Granular