missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ IKB zeta Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 64332 |
| Species | Mouse |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56390 |
| Gene Alias | FLJ30225, FLJ34463, ikappaBzeta, I-kappa-B-zeta, IkappaB-zeta, ikbzeta, IkB-zeta, IL-1 inducible nuclear ankyrin-repeat protein, INAPIkappa B-zeta variant 3, MAILIKBZikB-zeta, Molecule possessing ankyrin repeats induced by lipopolysaccharide, NF-kappa-B inhibitor zeta, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor |
| Gene Symbol | NFKBIZ |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Cell Cycle and Replication, Endocrinology, Immunology, Innate Immunity |
| Show More |