Learn More
Brady™ i7500 Printer Accessory Tear Plate for Peel Configuration Printer, Black
Description
- Optional serrated-edge plate can be added in place of the smooth-edge peel plate that ships standard on the Peel configuration printer. Intended for using a Peel configuration printer in non-peel (tear-off) mode with thicker liner materials that don't tear off with the smooth-edge peel plate. This tear plate is not the same as, nor is it interchangeable with the Standard configuration tear plate (#177729). Easily removed and installed using the included T-20 wrench that ships with all i7500 printer models.
Specifications
Specifications
| Product Type | Printer Plate |
| For Use With (Equipment) | i7500 |