Learn More
Eisco™ Igneous Rocks Kit with Storage Box and ID Card
A fun introduction to the world of geology through a 12-piece rock kit. Each Tech Cut specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features.
Supplier:  Eisco™ ESNGKIT0008
Catalog No. S012305
Only null left
Description
Description
  • Igneous rocks are formed when magma or lava cools and solidifies
  • Eisco Labs™ Tech Cut Rock and Mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
  • Set includes twelve different igneous rocks: Quartz Point, Andesite, Basalt, Tuff, Gabbro, White Granite, Pink Granite, Obsidian, Peridotite, Pumice, Rhyolite and Scoria
  • While each specimen will vary in shape and size, they will measure approximately 1 in. each
  • Test hardness, streak color, magnetic properties, appearance, and effervescence to identify the different specimens in this kit
  • Packaged in a box with substantial protective foam padding to store and preserve specimens
  • Ideal for home schooling or a fun home science activity, and is a great addition to any classroom or geology laboratory
  • Useful for demonstrations in identifying and classifying rocks and minerals, especially for group activities
Specifications
Specifications
Igneous Rock
Product Suggestions
Product Suggestions