missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IGH/MYC DY Translocation FISH Probe

Catalog No. 89999072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89999072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89999072 Supplier Abnova™ Supplier No. FT0026

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Labeled FISH probes for identification of gene translocation using Fluoresecent In Situ Hybridization Technique.

  • The IGH/MYC DY Translocation FISH Probe is designed to detect translocations involving the 14q32 and 8q24 chromosome regions.
  • Chromosome Region: 14q32/8q24
  • Fluorophore: FITC/Texas Red

Specifications

Content And Storage Store at 4°C in the dark.
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Detection Method Fluorescence
Format Tube
Label or Dye FITC
For Use With (Application) FISH-Ce
Gene ID (Entrez) 3492/4609
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Product Type FISH Probe
Quantity 100 μL
Target Human
Form Liquid
Show More Show Less