Recombinant Protein;Human IGFBP-7 Protein;50UG;Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession # NP_001544).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human IGFBP-7 Protein;50UG;Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession # NP_001544).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.