missing translation for 'onlineSavingsMsg'
Learn More

Murexide Indicator, 0.2% (w/w) in Sodium Chloride, Powder Mixture, For Calcium, APHA, Spectrum™ Chemical

Catalog No. 18611832
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18611832 100 g
18611833 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611832 Supplier Spectrum Chemical Mfg Cor Supplier No. M280100GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Murexide Indicator, 0.2 Percent (w/w) in Sodium Chloride, Powder Mixture, For Calcium, APHA
CAS 3051-09-0, 7647-14-5
Concentration 0.2 Percent (w/w)
Packaging Poly Bottle
Color Purple/Red to Red
Quantity 100 g
Physical Form Chemical Solution
Grade APHA

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.