missing translation for 'onlineSavingsMsg'
Learn More

Igepal(R) CO-520, Spectrum™ Chemical

Chemical Identifiers

CAS 26027-38-3
Molecular Weight (g/mol) 440.63
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611014
View Documents Promotion Details
N/A 20 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

26027-38-3
440.63

Specifications

Colorless, Viscous
1
440.63
Igepal CO to 520
26027-38-3
1.493-1.497
Ungraded
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.