missing translation for 'onlineSavingsMsg'
Learn More

Igepal(R) CA-720, Spectrum™ Chemical

Catalog No. 18611011
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611011 100 g Amber Glass Bottle
18611012 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611011 Supplier Spectrum Chemical Mfg Cor Supplier No. I1149100GM
Only null left

Chemical Identifiers

CAS 9036-19-5
Molecular Formula C18H30O3
Molecular Weight (g/mol) 294.44
InChI Key LBCZOTMMGHGTPH-UHFFFAOYSA-N
IUPAC Name 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1

Specifications

CAS Max % 1
pH Neutral
Quantity 100 g
Hydroxyl Value 0.5 mg/g
Packaging Amber Glass Bottle
Grade Ungraded
Chemical Name or Material Igepal CA to 720

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.