missing translation for 'onlineSavingsMsg'
Learn More

IGEPAL∣r CA-630

Catalog No. AAJ61055AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61055AE 100 mL
AAJ61055AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61055AE Supplier Thermo Scientific Chemicals Supplier No. J61055AE

IGEPAL∣r CA-630, CAS # 9002-93-1, is a nonionic and non-denaturing detergent used mainly for the solubilization, isolation, and purification of membrane protein complexes.

Detergent, equivalent to Nonidet P-40. For solubilizing membrane proteins, IGEPAL™ CA-630 is used as a detergent in paper industry, hand cleaners, metal cleaners. It is suitable for solubilization, isolation and purification of membrane protein complexes and in two-dimensional gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • IGEPAL|r CA-630 is a nonionic and non-denaturing detergent similar to Nonidet P-40

Application

  • IGEPAL|r CA-630 is mainly used for the solubilization, isolation, and purification of membrane protein complexes and in two-dimensional gel electrophoresis
  • IGEPAL|r CA-630 can be used in the composition of homogenizing buffers during western blotting technique in mouse muscle homogenates
  • It has been used in X-gal staining to study the lacZ expression
  • IGEPAL|r CA-630 is also a component of the cell lysis buffer in order to prepare RT-qPCR-ready cell lysates

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Octylphenyl-polyethylene glycol
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless
Physical Form Liquid
Density 1.060
Quantity 100 mL
Solubility Information Miscible with water.
Chemical Name or Material IGEPAL™ CA-630
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only