missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgA Rat ELISA Standard (for Uncoated ELISA Kit)

Catalog No. 501123567
Change view
Click to view available options
Quantity:
4 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501123567 4 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501123567 Supplier Invitrogen™ Supplier No. 395048065
Only null left

This product is a component of a respective eBioscience Kit. Please refer to the relevant manual for each product on our website.

This product is a component of a respective eBioscience Kit. Please refer to the relevant manual for each product on our website.

IgA comprises approximately 15% of all immunoglobulins in healthy human serum. IgA in serum is mainly monomeric, but in secretions, such as saliva, tears, colostrums, mucus, sweat, gastric fluid, IgA is found as a dimer where they are connected by a joining peptide. Most IgA is present in secreted form. This is believed to be due to ist properties in preventing invading pathogens by attaching and penetrating epithelial surfaces. IgA is just a very weak complement activating antibody; hence it does not induce bacterial cell lysis via the complement system. However secretory IgA works together with lysozymes, also present in many secreted fluids, which hydrolyze carbohydrates in bacterial cell walls enabling the immune system to clear the infection. IgA is predominantly found on epithelial cell surfaces where it acts as a neutralizing antibody.

Reported Application

  • ELISA, ELISA Recombinant Standard

Specifications

Immunoassay Kit Format ELISA (Standard)
Target Species Rat
Former Identifiers Ready-SET-Go!, eBioscience
For Use With (Equipment) Microplate Reader
Quantity 4 pack
Target Kit Named IgA
Target Species Validated Rat