missing translation for 'onlineSavingsMsg'
Learn More

Hemosure™ iFOB Test Kits

Catalog No. p-3111549
Change view
Click to view available options
Includes:
50 tests, 50 tubes, no mailers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23800512 50 tests, 50 tubes, no mailers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23800512 Supplier Hemosure™ Supplier No. T1CK50
Only null left

Qualitative, one-step, odorless iFOB test with high sensitivity and specificity

  • CLIA Complexity: Waived
  • CPT Codes: G0328QW (colorectal cancer screening); 82274QW (detection of blood in feces form)
  • Sensitivity: 50ng hHB/mL
  • Specificity: over 96%
  • Useful aid in the diagnosis and therapy of gastrointestinal disorders
  • Patient-friendly for use in physicians' offices, labs, hospitals, clinics, or homes
  • As test sample flows up through absorbent device, the labeled antibody-dye conjugate binds to the hemoglobin in specimen, forming an antibody-antigen complex
  • Results within five minutes
  • Positive results: one band in the control (C) region, one band in the test (T) region
  • Negative results: only one band in the control (C) region
  • Low false positive results
  • Reaction components: citric acid, monoclonal and polyclonal antibody, mouse anti-HB antibodies 1 and 2, and goat anti-mouse IgG antibody

If home kit mailers are not needed, they can be excluded from the order. Please contact your Fisher HealthCare account manager for details.

Order Info

The 10-test kit contains 10 collection bottles and 10 home kit mailers; 30-test kit contains 50 collection bottles and 50 home kit mailers.

Specifications

Content And Storage 50 Fecal occult Blood test cassettes, 50 collection tubes
CPT Code G0328QW, 82274QW
Clia Complexity Waived
Type iFOB Test Kit
For Use With (Application) Diagnosis and therapy of gastrointestinal disorders
Includes 50 tests, 50 tubes, no mailers
Quantity 50 Pack
Test Time 5 min.