missing translation for 'onlineSavingsMsg'
Learn More

Innovatek iFOB TEST™ Immunochemical Rapid Test

Catalog No. 361044576
Change view
Click to view available options
Quantity:
20 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361044576 20 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361044576 Supplier Innovatek Supplier No. 4194IFOB20

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

iFOB TEST™ Immunochemical Rapid Test detects the presence of occult blood in stool

  • Immunochemical method
  • Highest degree of sensitivity and specificity
  • One step procedure
  • No patient diet or medication restrictions
  • Convenient package size of 20 tests per kit
  • Includes test cassettes, collection device, and collection paper.

Specifications

Detectable Analytes Fecal Occult Blood
Sensitivity Highest Degree
Disposable Single-use
For Use With (Application) Early Colorectal Cancer Detection
Includes Test Cassettes, Collection Device, and Collection Paper
Quantity 20 Tests/Kit
Specificity Highest Degree