missing translation for 'onlineSavingsMsg'
Learn More

Jaece Industries Identi-Marker Pens

Catalog No. 14127112
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14127112 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14127112 Supplier Jaece Industries Supplier No. IMF200
Only null left

Writes on foam, plastic, glass and metal. Jaece Identi-Marker Pens are ideal for use with Identi-Plugs™ and other Jaece products.

  • Ideal for use with Identi-Plugs™ and other Jaece products
  • Writes on cold or wet surfaces
  • Marks on plastic, glass and metal
  • Permanent, quick-drying ink
  • Extra-fine, durable plastic tip
  • Ideal for general laboratory, hospital or industrial use

Specifications

Product Type Identi-Marker Pen
Color Blue
For Use With (Application) General laboratory, hospital or industrial use
For Use With (Equipment) Identi-Plugs™ and other Jaece products