missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-MS Calibration Standard 1, Method 200.8 (91), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | ICP-MS Calibration Standard 1 |
| Packaging | Natural LDPE Bottle |
| CAS Min % | 89.0, 7.23, 0.01, 0.01, 0.01, 0.0023 |
| CAS Max % | 92.0, 7.53, 0.01, 0.01, 0.01, 0.0023 |
| CAS | 7697-37-2, 7784-27-2, 7789-02-8, 7647-01-0, 13823-29-5 |
| Odor | Odorless |