Learn More
Dentec Safety IceTred™ FSTD - Full Sole Traction Devices
Catalog No. 19999687
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19999687 Small
19999688 Medium
19999689 Large
19999686 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19999687 Supplier Dentec Safety Supplier No. 10710 S
Only null left
IceTred™ FSTD - Full Sole Traction Devices - Run or walk on ice or snow with confidence.
  • Tough and Durable Design - Made of durable natural rubber that is incredibly tough but maintains its flexibility in extreme temperatures. Extra thick toe cap straps make it easy to fit all footwear styles
  • Tiny Solutions BIG GRIP - Six strategically positioned tungsten carbide spikes provide excellent ice traction performance while maintaining comfort under foot. Wears longer and holds its ice piercing edge better than steel spikes or screw heads. Becomes sharper with greater wear. Spark resistant. Used on all ICETRED™ Traction Devices. Rubber Traction studs provide excellent traction on snow
  • Easy to use - Extra large heel tab strap makes it easy to pull the heel pad over any footwear
  • Not a Spark Hazard - Third party tested not to be a spark hazard in gas groups A – D as per CSA C22.2 No. 213-17 combustibility testing protocol. If you require intrinsic safety protection from static discharge, must use approved Electrostatic Discharging (ESD) footwear or clothing and/or accessories that offer ESD protection with our ICETRED™ traction devices
Specifications
Size Small
Product Type Safety Soles