Learn More
Dentec Safety IceTred™ FSTD - Full Sole Traction Devices
IceTred™ FSTD - Full Sole Traction Devices - Run or walk on ice or snow with confidence.
Supplier: Dentec Safety 10710XL
Description
- Tough and Durable Design - Made of durable natural rubber that is incredibly tough but maintains its flexibility in extreme temperatures. Extra thick toe cap straps make it easy to fit all footwear styles
- Tiny Solutions BIG GRIP - Six strategically positioned tungsten carbide spikes provide excellent ice traction performance while maintaining comfort under foot. Wears longer and holds its ice piercing edge better than steel spikes or screw heads. Becomes sharper with greater wear. Spark resistant. Used on all ICETRED™ Traction Devices. Rubber Traction studs provide excellent traction on snow
- Easy to use - Extra large heel tab strap makes it easy to pull the heel pad over any footwear
- Not a Spark Hazard - Third party tested not to be a spark hazard in gas groups A – D as per CSA C22.2 No. 213-17 combustibility testing protocol. If you require intrinsic safety protection from static discharge, must use approved Electrostatic Discharging (ESD) footwear or clothing and/or accessories that offer ESD protection with our ICETRED™ traction devices
Specifications
| X-Large | |
| Safety Soles |