Learn More
Ice Melting Set
Catalog No. S68112
Change view
Click to view available options
Product Type:
Ice Melting Set
Catalog No. Product Type
S68112 Ice Melting Set
1 options
Catalog No. S68112 Supplier Customer Channels Group (Ccg) Supplier No. 15057
Includes
Instructions with theory
Two black blocks simply and quickly demonstrate differences in heat conductivity.
  • One block is made from aluminum, which is an excellent conductor of heat
  • Other is made from plastic foam, which is a poor conductor of heat
  • Simply put an ice cube on each block and watch as one quickly melts away
  • Rim on each block prevents spillage
Specifications
For Use With (Application) To demonstrate differences in heat conductivity
Includes Aluminum base and plastic foam base, both with ring to prevent spillage, instructions
Product Type Ice Melting Set