missing translation for 'onlineSavingsMsg'
Learn More

Icariin, 98%

Catalog No. aaj66324md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66324MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66324MD Supplier Thermo Scientific Chemicals Supplier No. J66324MD

CAS: 489-32-7 | C33H40O15 | 676.668 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 489-32-7
Color Yellow
Molecular Formula C33H40O15
Quantity 250 mg
MDL Number MFCD00210516
Merck Index 14,3617
Synonym 3-[(6-Deoxy-alpha-L-mannopyranosyl)oxy]-7-beta-D-glucopyranosyloxy-5-hydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one
InChI Key TZJALUIVHRYQQB-JOJMVVPMSA-N
SMILES CC1C(C(C(C(O1)OC2=C(OC3=C(C2=O)C(=CC(=C3CC=C(C)C)OC4C(C(C(C(O4)CO)O)O)O)O)C5=CC=C(C=C5)OC)O)O)O
IUPAC Name 5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one
Molecular Weight (g/mol) 676.668
PubChem CID 131801625
Formula Weight 676.66
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Icariin
Show More Show Less
RTECSNumber DJ2980500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only