Learn More
Waters Limited IC-Pak Cation M/D Column, 3.9 mm x 150 mm, 1/pk
Catalog No. 50818784
Encompass
The silica-based IC-Pak C M/D column allows a simultaneous analysis of monovalent and divalent cations under isocratic conditions. This technology provides the analytical chemist with the most sensitive means of analyzing monovalent cations such as Na+, K+, Li+ and NH4+.
Catalog No. 50818784 Supplier Waters Limited Supplier No. WAT036570
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The silica-based IC-Pak C M/D column allows a simultaneous analysis of monovalent and divalent cations under isocratic conditions. This technology provides the analytical chemist with the most sensitive means of analyzing monovalent cations such as Na+, K+, Li+ and NH4+.