Learn More
Waters Limited IC-Pak Anion-Exchange Guard-Pak Insert, 10 m, 5/pk
Catalog No. 50784973
Encompass
Waters offers an array of products for ion chromatography . IC-Pak resin based columns allow you to analyze a full range of ions from numerous sample matrices, both simple and complex. They offer an exceptional linear loading range of less than 1.0 ppb to greater than 400 ppm without dilution and without pH limitation on eluent or sample. The flexibility exists for accurate and reproducible anion and cation analyses at all concentration levels.
Catalog No. 50784973 Supplier Waters Limited Supplier No. WAT010551
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Waters offers an array of products for ion chromatography . IC-Pak resin based columns allow you to analyze a full range of ions from numerous sample matrices, both simple and complex. They offer an exceptional linear loading range of less than 1.0 ppb to greater than 400 ppm without dilution and without pH limitation on eluent or sample. The flexibility exists for accurate and reproducible anion and cation analyses at all concentration levels.